siRNA Details
Gene Symbol: Slu7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100550.1 | NP_001094020.1 | 3 | 379 | ||
XM_008767640.1 | XP_008765862.1 | 2 | 1154 | |||
XM_008767641.1 | XP_008765863.1 | 3 | 826 | |||
XM_008767642.1 | XP_008765864.1 | 3 | 1031 | |||
GenBank mRNA | BC062243.1 | - | 3 | 383 | ||
BC158865.1 | - | 3 | 345 | 7938574 |
