siRNA Details
Gene Symbol: Ulk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191645.1 | NP_001178574.1 | 16, 17 | 1960 | ||
XM_006246497.2 | XP_006246559.1 | 16, 17 | 1941 | |||
XM_006246498.2 | XP_006246560.1 | 16, 17 | 1940 | |||
XM_006246499.2 | XP_006246561.1 | 15, 16 | 1804 |
