siRNA Details
Gene Symbol: Cntrob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134645.1 | NP_001128117.1 | 18, 19 | 2786 | ||
XM_006246701.2 | XP_006246763.1 | 19, 20 | 3160 | |||
XM_008767825.1 | XP_008766047.1 | 18, 19 | 2494 | |||
GenBank mRNA | EF532342.1 | - | 18, 19 | 2786 | ||
EF532343.1 | - | 18, 19 | 2838 | |||
EF532344.1 | - | 18, 19 | 2845 | |||
EF532345.1 | - | 18, 19 | 2838 | |||
EF532346.1 | - | 17, 18 | 3108 | |||
EF532347.1 | - | 18, 19 | 3156 | |||
EF532348.1 | - | 19, 20 | 3182 | |||
EF532349.1 | - | 19, 20 | 3214 | |||
EF532350.1 | - | 17, 18 | 3192 |
