siRNA Details
Gene Symbol: Cntrob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134645.1 | NP_001128117.1 | 17 | 2631 | ||
XM_006246701.2 | XP_006246763.1 | 18 | 3005 | |||
XM_008767825.1 | XP_008766047.1 | 17 | 2339 | |||
GenBank mRNA | EF532342.1 | - | 17 | 2631 | ||
EF532343.1 | - | 17 | 2683 | |||
EF532344.1 | - | 17 | 2690 | |||
EF532345.1 | - | 17 | 2683 | |||
EF532346.1 | - | 16 | 2953 | |||
EF532347.1 | - | 17 | 3001 | |||
EF532348.1 | - | 18 | 3027 | |||
EF532349.1 | - | 18 | 3059 | |||
EF532350.1 | - | 16 | 3037 |
