siRNA Details
Gene Symbol: Cntrob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134645.1 | NP_001128117.1 | 13 | 2147 | ||
XM_006246701.2 | XP_006246763.1 | 14 | 2521 | |||
XM_006246702.2 | XP_006246764.1 | 13 | 2795 | |||
XM_008767825.1 | XP_008766047.1 | 13 | 1855 | |||
GenBank mRNA | EF532342.1 | - | 13 | 2147 | ||
EF532343.1 | - | 13 | 2199 | |||
EF532344.1 | - | 13 | 2206 | |||
EF532345.1 | - | 13 | 2199 | |||
EF532346.1 | - | 12 | 2469 | |||
EF532347.1 | - | 13 | 2517 | |||
EF532348.1 | - | 14 | 2543 | |||
EF532349.1 | - | 14 | 2575 | |||
EF532350.1 | - | 12 | 2553 |
