siRNA Details
Gene Symbol: Dnah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767898.1 | XP_008766120.1 | 35, 36 | 5966 | ||
XM_008767899.1 | XP_008766121.1 | 35, 36 | 5802 | |||
XM_008767900.1 | XP_008766122.1 | 29, 30 | 4765 | |||
GenBank mRNA | AY903242.1 | - | 24, 25 | 3865 | ||
AY903243.1 | - | 24, 25 | 3865 | |||
AY903244.1 | - | 24, 25 | 3865 |
