siRNA Details
Gene Symbol: Senp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013116.2 | NP_001013134.2 | 11 | 1835 | ||
XM_008767826.1 | XP_008766048.1 | 11 | 1973 | |||
GenBank mRNA | BC101901.1 | - | 10 | 1660 |
