siRNA Details
Gene Symbol: Senp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013116.2 | NP_001013134.2 | 5 | 1296 | ||
XM_006246703.2 | XP_006246765.1 | 5 | 1345 | |||
XM_008767826.1 | XP_008766048.1 | 5 | 1434 | |||
GenBank mRNA | BC090032.1 | - | 1 | 426 | ||
BC101901.1 | - | 4 | 1121 |
