siRNA Details
Gene Symbol: Tnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107012.1 | NP_001100482.1 | 6 | 1032 | ||
XM_006246704.2 | XP_006246766.1 | 7 | 2490 | |||
XM_006246705.2 | XP_006246767.1 | 8 | 1565 | |||
XM_006246707.2 | XP_006246769.1 | 8 | 1235 | |||
XM_006246709.2 | XP_006246771.1 | 7 | 1412 | |||
XM_008767827.1 | XP_008766049.1 | 7 | 1941 | |||
XM_008767828.1 | XP_008766050.1 | 8 | 1329 |
