siRNA Details
Gene Symbol: Fbxo39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039018.1 | NP_001034107.1 | 1 | 1013 | ||
XM_006246727.2 | XP_006246789.1 | 2 | 1270 | |||
XM_006246729.2 | XP_006246791.1 | 2 | 1110 | |||
GenBank mRNA | BC082089.1 | - | 2 | 1063 | ||
BC100079.1 | - | 1 | 1013 | 7132945 |
