siRNA Details
Gene Symbol: Cluh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246858.2 | XP_006246920.1 | 10 | 4978 | ||
XM_006246859.2 | XP_006246921.1 | 10 | 4978 | |||
XM_006246860.2 | XP_006246922.1 | 10 | 4963 | |||
XM_006246861.2 | XP_006246923.1 | 10 | 4963 | |||
XM_006246862.2 | XP_006246924.1 | 10 | 1763 | |||
XM_006246864.2 | XP_006246926.1 | 11 | 1798 | |||
XM_008767914.1 | XP_008766136.1 | 10 | 1813 | |||
XM_008767915.1 | XP_008766137.1 | 10 | 1873 | |||
XM_008767916.1 | XP_008766138.1 | 11 | 1511 | |||
XM_220693.9 | XP_220693.6 | 10 | 1746 |
