siRNA Details
Gene Symbol: LOC103694864 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773811.1 | XP_008772033.1 | 6 | 1145 |
Gene Symbol: Mmp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079888.1 | NP_001073357.1 | 6 | 954 | ||
GenBank mRNA | EF121988.1 | - | 6 | 954 | ||
EF121989.1 | - | 6 | 954 |
