siRNA Details
Gene Symbol: Appbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100969.1 | NP_001094439.1 | 3 | 563 | ||
GenBank mRNA | BC091285.1 | - | 2 | 137 | ||
BC169030.1 | - | 3 | 564 | 7446141 | ||
EF535262.1 | - | 3 | 401 |
