siRNA Details
Gene Symbol: Rnf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135921.1 | NP_001129393.1 | 8 | 2015 | ||
XM_006247090.2 | XP_006247152.1 | 9 | 3500 | |||
XM_006247091.2 | XP_006247153.1 | 9 | 3275 | |||
XM_006247092.2 | XP_006247154.1 | 8 | 4020 | |||
XM_006247094.2 | XP_006247156.1 | 7 | 3376 | |||
XM_008768038.1 | XP_008766260.1 | 8 | 4411 | |||
XM_008768039.1 | XP_008766261.1 | 6 | 4054 | |||
XM_008768040.1 | XP_008766262.1 | 7 | 3956 | |||
XM_008768041.1 | XP_008766263.1 | 7 | 3604 |
