siRNA Details
Gene Symbol: Dnajc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213625.1 | NP_998790.1 | 4 | 312 | ||
XM_006247311.2 | XP_006247373.1 | 5 | 673 | |||
XM_006247312.2 | XP_006247374.1 | 4 | 393 | |||
XM_006247313.1 | XP_006247375.1 | 3 | 233 | |||
GenBank mRNA | AB178470.1 | - | 4 | 312 | ||
BC093600.1 | - | 3 | 228 | 7316009 | ||
FQ212948.1 | - | 4 | 343 |
