siRNA Details
Gene Symbol: Ezh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107051.1 | NP_001100521.1 | 9 | 1077 | ||
XM_006247318.2 | XP_006247380.1 | 10 | 1123 | |||
XM_006247319.2 | XP_006247381.1 | 10 | 1187 | |||
XM_008768052.1 | XP_008766274.1 | 10 | 1128 |
