siRNA Details
Gene Symbol: Ezh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107051.1 | NP_001100521.1 | 7, 8 | 830 | ||
XM_006247318.2 | XP_006247380.1 | 8, 9 | 876 | |||
XM_006247319.2 | XP_006247381.1 | 8, 9 | 940 | |||
XM_008768052.1 | XP_008766274.1 | 8, 9 | 881 |

More Information
Other Species Targeted

Gene Ontology Categories:
