siRNA Details
Gene Symbol: Plekhm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009677.1 | NP_001009677.1 | 4 | 879 | ||
XM_006247527.2 | XP_006247589.1 | 4 | 919 | |||
XM_008768292.1 | XP_008766514.1 | 3 | 699 | |||
GenBank mRNA | BC087058.1 | - | 4 | 879 | 7124252 |
