siRNA Details
Gene Symbol: Tanc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191653.1 | NP_001178582.1 | 25, 26 | 4300 | ||
XM_008768351.1 | XP_008766573.1 | 27, 28 | 4449 | |||
XM_008768352.1 | XP_008766574.1 | 27, 28 | 4439 | |||
XM_008768353.1 | XP_008766575.1 | 26, 27 | 4419 | |||
XM_008768354.1 | XP_008766576.1 | 26, 27 | 4338 | |||
XM_008768355.1 | XP_008766577.1 | 27, 28 | 4338 | |||
XM_008768356.1 | XP_008766578.1 | 26, 27 | 4227 |
