siRNA Details
Gene Symbol: Arsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047877.1 | NP_001041342.1 | 3, 4 | 452 | ||
XM_006247599.2 | XP_006247661.1 | 5, 6 | 2652 | |||
XM_006247600.2 | XP_006247662.1 | 4, 5 | 2509 | |||
XM_006247601.2 | XP_006247663.1 | 6, 7 | 2878 | |||
XM_006247602.2 | XP_006247664.1 | 5, 6 | 1440 | |||
XM_006247603.2 | XP_006247665.1 | 5, 6 | 1294 | |||
XM_006247604.2 | XP_006247666.1 | 5, 6 | 2710 | |||
XM_006247606.2 | XP_006247668.1 | 4, 5 | 2567 | |||
XM_006247607.2 | XP_006247669.1 | 4, 5 | 1479 | |||
XM_006247608.2 | XP_006247670.1 | 3, 4 | 1129 | |||
XM_006247609.2 | XP_006247671.1 | 3, 4 | 424 | |||
XM_008768368.1 | XP_008766590.1 | 6, 7 | 2821 | |||
XM_008768369.1 | XP_008766591.1 | 7, 8 | 2285 |
