siRNA Details
Gene Symbol: Rhbdf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107067.2 | NP_001100537.2 | 19 | 2449 | ||
NM_001270834.1 | NP_001257763.1 | 19 | 2377 | |||
GenBank mRNA | FQ222541.1 | - | 19 | 2377 | ||
FQ230904.1 | - | 19 | 2449 |
