siRNA Details
Gene Symbol: Cd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107074.1 | NP_001100544.1 | 2 | 178 | ||
GenBank mRNA | BC167030.1 | - | 2 | 150 | 7378211 | |
FQ226313.1 | - | 2 | 180 | |||
FQ226643.1 | - | 2 | 180 | |||
FQ227263.1 | - | 2 | 180 |
