siRNA Details
Gene Symbol: Slc7a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107078.2 | NP_001100548.1 | 3 | 1505 | ||
XM_006248693.2 | XP_006248755.1 | 4 | 1878 | |||
XM_006248694.2 | XP_006248756.1 | 4 | 1698 | |||
XM_006248695.2 | XP_006248757.1 | 4 | 1875 | |||
XM_006248696.2 | XP_006248758.1 | 3 | 1733 | |||
XM_006248697.2 | XP_006248759.1 | 3 | 1730 | |||
XM_006248698.2 | XP_006248760.1 | 3 | 1582 | |||
GenBank mRNA | BC169079.1 | - | 3 | 1505 | 7937326 | |
FQ214252.1 | - | 3 | 1699 |
