siRNA Details
Gene Symbol: Bcl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107084.1 | NP_001100554.1 | 5 | 772 | ||
XM_008768799.1 | XP_008767021.1 | 4 | 1645 | |||
GenBank mRNA | BC166425.1 | - | 5 | 672 | 7097735 |
