siRNA Details
Gene Symbol: Parp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191659.1 | NP_001178588.1 | 14 | 4546 | ||
GenBank mRNA | FQ218194.1 | - | 7 | 1165 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Parp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191659.1 | NP_001178588.1 | 14 | 4546 | ||
GenBank mRNA | FQ218194.1 | - | 7 | 1165 |