siRNA Details
Gene Symbol: Bbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079938.1 | NP_001073407.1 | 13 | 2558 | ||
XM_008768634.1 | XP_008766856.1 | 14 | 2540 | |||
XM_008768635.1 | XP_008766857.1 | 14 | 2735 | |||
XM_008768636.1 | XP_008766858.1 | 15 | 2849 | |||
XM_008768637.1 | XP_008766859.1 | 15 | 2654 | |||
XM_008768638.1 | XP_008766860.1 | 13 | 2450 | |||
XM_008768639.1 | XP_008766861.1 | 13 | 2645 | |||
XM_008768640.1 | XP_008766862.1 | 14 | 2426 | |||
XM_008768641.1 | XP_008766863.1 | 12 | 2390 | |||
GenBank mRNA | BC129113.1 | - | 13 | 2558 | 7112122 |

More Information
Other Species Targeted

Gene Ontology Categories:
