siRNA Details
Gene Symbol: Il10rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107111.1 | NP_001100581.1 | 6 | 776 | ||
XM_006248062.2 | XP_006248124.1 | 6 | 1008 | |||
XM_006248063.2 | XP_006248125.1 | 6 | 1352 |
