siRNA Details
Gene Symbol: Arhgef18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107115.1 | NP_001100585.1 | 17 | 2350 | ||
XM_006248760.2 | XP_006248822.1 | 18 | 2308 | |||
XM_008768950.1 | XP_008767172.1 | 16 | 2349 | |||
XM_008768951.1 | XP_008767173.1 | 12 | 1501 | |||
XM_008768952.1 | XP_008767174.1 | 16 | 1866 |
