siRNA Details
Gene Symbol: Fry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170398.1 | NP_001163869.1 | 37 | 5293 | ||
XM_008768960.1 | XP_008767182.1 | 38 | 6568 | |||
GenBank mRNA | EU563850.1 | - | 37 | 5293 | ||
EU563851.1 | - | 37 | 5293 |
