siRNA Details
Gene Symbol: Fbxo24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107130.2 | NP_001100600.2 | 8 | 1221 | ||
XM_006249139.2 | XP_006249201.1 | 8 | 1734 | |||
XM_008769135.1 | XP_008767357.1 | 8 | 1803 | |||
XM_008769136.1 | XP_008767358.1 | 9 | 1825 |
