siRNA Details
Gene Symbol: Glt1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769205.1 | XP_008767427.1 | 10, 11 | 1047 | ||
XM_222147.8 | XP_222147.4 | 10, 11 | 902 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Glt1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769205.1 | XP_008767427.1 | 10, 11 | 1047 | ||
XM_222147.8 | XP_222147.4 | 10, 11 | 902 |