siRNA Details
Gene Symbol: Oas1i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009680.1 | NP_001009680.1 | 4 | 848 | ||
GenBank mRNA | AY196697.1 | - | 4 | 848 | ||
BC160819.1 | - | 4 | 822 | 7382468 |
Gene Symbol: LOC100910735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751197.2 | XP_003751245.1 | 868 |
