siRNA Details
Gene Symbol: Oas1i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009680.1 | NP_001009680.1 | 1 | 114 | ||
GenBank mRNA | AY196697.1 | - | 1 | 114 | ||
BC160819.1 | - | 1 | 88 | 7382468 |
Gene Symbol: LOC100910735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751197.2 | XP_003751245.1 | 134 |
