siRNA Details
Gene Symbol: Ep400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107149.1 | NP_001100619.1 | 33 | 6307 | ||
XM_006249549.2 | XP_006249611.1 | 35 | 6661 | |||
XM_006249550.2 | XP_006249612.1 | 35 | 6826 | |||
XM_006249552.2 | XP_006249614.1 | 35 | 6642 | |||
XM_006249553.2 | XP_006249615.1 | 35 | 6662 | |||
XM_006249554.2 | XP_006249616.1 | 35 | 6661 | |||
XM_006249556.2 | XP_006249618.1 | 34 | 6553 | |||
XM_006249557.2 | XP_006249619.1 | 34 | 6550 | |||
XM_006249558.2 | XP_006249620.1 | 34 | 6418 | |||
XM_006249559.2 | XP_006249621.1 | 32 | 6198 | |||
XM_006249560.2 | XP_006249622.1 | 34 | 6549 | |||
XM_008769332.1 | XP_008767554.1 | 36 | 6763 |
