siRNA Details
Gene Symbol: Sart3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107156.1 | NP_001100626.1 | 7, 8 | 1079 | ||
XM_006249501.2 | XP_006249563.1 | 7, 8 | 1452 | |||
XM_006249502.2 | XP_006249564.1 | 8, 9 | 1084 | |||
XM_008769338.1 | XP_008767560.1 | 7, 8 | 1892 |
