siRNA Details
Gene Symbol: Serpinb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107168.1 | NP_001100638.1 | 8 | 930 | ||
XM_008769454.1 | XP_008767676.1 | 8 | 930 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Serpinb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107168.1 | NP_001100638.1 | 8 | 930 | ||
XM_008769454.1 | XP_008767676.1 | 8 | 930 |