siRNA Details
Gene Symbol: Zranb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249701.2 | XP_006249763.1 | 8, 9 | 1132 | ||
XM_006249702.2 | XP_006249764.1 | 8, 9 | 1779 | |||
XM_006249704.1 | XP_006249766.1 | 8, 9 | 1693 |
