siRNA Details
Gene Symbol: Kdm5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107177.1 | NP_001100647.1 | 14 | 2165 | ||
XM_008769555.1 | XP_008767777.1 | 14 | 2201 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kdm5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107177.1 | NP_001100647.1 | 14 | 2165 | ||
XM_008769555.1 | XP_008767777.1 | 14 | 2201 |