siRNA Details
Gene Symbol: Ipo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107180.1 | NP_001100650.1 | 14 | 1603 | ||
XM_008769563.1 | XP_008767785.1 | 15 | 1595 |

More Information
Other Species Targeted

Gene Ontology Categories:
