siRNA Details
Gene Symbol: Rnf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025667.1 | NP_001020838.1 | 5 | 871 | ||
XM_006249990.2 | XP_006250052.1 | 5 | 809 | |||
XM_006249991.2 | XP_006250053.1 | 5 | 891 | |||
GenBank mRNA | AB568264.1 | - | 4 | 675 | ||
BC098941.1 | - | 5 | 871 | 7456372 |
