siRNA Details
Gene Symbol: Dhx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107184.1 | NP_001100654.1 | 15 | 1815 | ||
XM_006249995.2 | XP_006250057.1 | 15 | 1851 | |||
XM_006249996.2 | XP_006250058.1 | 15 | 1848 | |||
XM_008769633.1 | XP_008767855.1 | 10 | 1121 |
