siRNA Details
Gene Symbol: Tbx19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107193.1 | NP_001100663.1 | 2 | 501 | ||
XM_008769651.1 | XP_008767873.1 | 2 | 1177 | |||
XM_008769652.1 | XP_008767874.1 | 3 | 538 |
