siRNA Details
Gene Symbol: Atf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107196.1 | NP_001100666.1 | 4, 5 | 391 | ||
XM_008769738.1 | XP_008767960.1 | 4, 5 | 447 | |||
GenBank mRNA | BC168890.1 | - | 4, 5 | 374 | 7108280 |
