siRNA Details
Gene Symbol: Fbxo28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107203.3 | NP_001100673.1 | 4 | 600 | ||
GenBank mRNA | BC167003.1 | - | 4 | 601 | 8366732 | |
FQ223120.1 | - | 3 | 296 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fbxo28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107203.3 | NP_001100673.1 | 4 | 600 | ||
GenBank mRNA | BC167003.1 | - | 4 | 601 | 8366732 | |
FQ223120.1 | - | 3 | 296 |