siRNA Details
Gene Symbol: Fbxo28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107203.3 | NP_001100673.1 | 5 | 790 | ||
GenBank mRNA | BC167003.1 | - | 5 | 791 | 8366732 |

More Information
Other Species Targeted

