siRNA Details
Gene Symbol: Zfp644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139484.1 | NP_001132956.1 | 7 | 3568 | ||
XM_006250550.1 | XP_006250612.1 | 2 | 3255 | |||
XM_006250554.2 | XP_006250616.1 | 2 | 3265 | |||
XM_008769931.1 | XP_008768153.1 | 2 | 3329 | |||
XM_008769932.1 | XP_008768154.1 | 3 | 3159 | |||
XM_008769933.1 | XP_008768155.1 | 2 | 3255 | |||
XM_008769934.1 | XP_008768156.1 | 3 | 3233 | |||
XM_008769935.1 | XP_008768157.1 | 2 | 3255 |
