siRNA Details
Gene Symbol: Aff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107206.1 | NP_001100676.1 | 14, 15 | 3049 | ||
XM_006250632.2 | XP_006250694.1 | 15, 16 | 3154 | |||
XM_006250633.2 | XP_006250695.1 | 14, 15 | 3038 |
