siRNA Details
Gene Symbol: Adamts3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107212.1 | NP_001100682.1 | 13 | 2245 | ||
XM_006250773.2 | XP_006250835.1 | 13 | 2245 | |||
XM_006250774.2 | XP_006250836.1 | 13 | 1870 |
