siRNA Details
Gene Symbol: Cep135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250874.2 | XP_006250936.1 | 19 | 2696 | ||
XM_006250875.2 | XP_006250937.1 | 19 | 4386 | |||
XM_223341.8 | XP_223341.6 | 19 | 2572 |
