siRNA Details
Gene Symbol: Slain2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107214.1 | NP_001100684.1 | 4 | 989 | ||
XM_006250898.2 | XP_006250960.1 | 4 | 1061 | |||
XM_006250899.2 | XP_006250961.1 | 4 | 1060 | |||
GenBank mRNA | BC158815.1 | - | 2 | 62 |
